Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzaldehyde, 98+%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
456-48-4
C7H5FO
124.11
MFCD00003348
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
m-fluorobenzaldehyde,benzaldehyde, 3-fluoro,3'-fluoro-benzaldehyde,3-fluoro-benzaldehyde,meta-fluorobenzaldehyde,benzaldehyde, m-fluoro,3-fc6h4cho,3-fluorobenzaldehyde,3-fluoro benzaldehyde,pubchem3477
68009
3-fluorobenzaldehyde
C1=CC(=CC(=C1)F)C=O
456-48-4
C7H5FO
124.11
MFCD00003348
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
m-fluorobenzaldehyde,benzaldehyde, 3-fluoro,3'-fluoro-benzaldehyde,3-fluoro-benzaldehyde,meta-fluorobenzaldehyde,benzaldehyde, m-fluoro,3-fc6h4cho,3-fluorobenzaldehyde,3-fluoro benzaldehyde,pubchem3477
68009
3-fluorobenzaldehyde
C1=CC(=CC(=C1)F)C=O
Density 1.1700g/mL
Refractive Index 1.5170 to 1.519
Boiling Point 66°C to 68°C (20.0mmHg)
Flash Point 56°C
Specific Gravity 1.17
Color Colorless to Yellow
Quantity 50mL
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula FC6H4CHO
Beilstein 07, II, 177
Formula Weight 124.11
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 3-Fluorobenzaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning