Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzamide, 99%, Thermo Scientific™

Catalog Number AC184080100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
455-37-8
C7H6FNO
139.13
MFCD00007983
YPIGHNIIXYSPKF-UHFFFAOYSA-N
m-fluorobenzamide, benzamide, 3-fluoro, 3-fluoro-benzamide, benzamide, m-fluoro, 3-fluoranylbenzamide, 3-fluorobenzamide, ksc492m7t, ypighniixyspkf-uhfffaoysa
68000
3-fluorobenzamide
NC(=O)C1=CC=CC(F)=C1
455-37-8
C7H6FNO
139.13
MFCD00007983
YPIGHNIIXYSPKF-UHFFFAOYSA-N
m-fluorobenzamide, benzamide, 3-fluoro, 3-fluoro-benzamide, benzamide, m-fluoro, 3-fluoranylbenzamide, 3-fluorobenzamide, ksc492m7t, ypighniixyspkf-uhfffaoysa
68000
3-fluorobenzamide
NC(=O)C1=CC=CC(F)=C1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 129°C to 132°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4CONH2
Quantity 10 g
Beilstein 09, I, 137
Formula Weight 139.13
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Fluorobenzamide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 207-247-5

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit