Alfa Aesar™ 3-Fluorobenzenesulfonamide, 97%

Manufacturer: Alfa Aesar™ H55740MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55740MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1524-40-9
    Formula Weight 175.18
    MDL Number MFCD00042284
    UN Number UN2811
    Chemical Name or Material 3-Fluorobenzenesulfonamide

    Specifications

    Formula Weight 175.18
    Melting Point 124° to 129°C
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00042284
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder
    CAS 1524-40-9
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 250mg
    UN Number UN2811
    Health Hazard 1 Danger
    Color White to brown
    Chemical Name or Material 3-Fluorobenzenesulfonamide
    Assay 97%

    Description

    Applications
    It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 

    Solubility
    Slightly soluble in water.

    Notes
    Store at room temperature. Avoid strong oxidizing agents.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit