Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
701-27-9
C6H4ClFO2S
194.60
MFCD00042286
OKYSUJVCDXZGKE-UHFFFAOYSA-N
3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
2734258
3-fluorobenzene-1-sulfonyl chloride
FC1=CC(=CC=C1)S(Cl)(=O)=O
701-27-9
C6H4ClFO2S
194.60
MFCD00042286
OKYSUJVCDXZGKE-UHFFFAOYSA-N
3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
2734258
3-fluorobenzene-1-sulfonyl chloride
FC1=CC(=CC=C1)S(Cl)(=O)=O
Refractive Index 1.5280 to 1.5310
Infrared Spectrum Authentic
Color Brown to Brown-Gray
Quantity 1g
Packaging Glass bottle
Boiling Point 108.0°C (15.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H4SO2Cl
Formula Weight 194.61
Physical Form Powder, Granules or Chunks
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzenesulfonyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger