Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
455-38-9
C7H5FO2
140.11
MFCD00002489
MXNBDFWNYRNIBH-UHFFFAOYSA-N
m-fluorobenzoic acid,benzoic acid, 3-fluoro,meta-fluorobenzoic acid,benzoic acid, m-fluoro,3-fluoro-benzoic acid,3-fluorobenzoicacid,pubchem3480,3-fluorobenzoic acid,ksc235q1b,paragos 390192
9968
CHEBI:20021
3-fluorobenzoic acid
C1=CC(=CC(=C1)F)C(=O)O
455-38-9
C7H5FO2
140.11
MFCD00002489
MXNBDFWNYRNIBH-UHFFFAOYSA-N
m-fluorobenzoic acid,benzoic acid, 3-fluoro,meta-fluorobenzoic acid,benzoic acid, m-fluoro,3-fluoro-benzoic acid,3-fluorobenzoicacid,pubchem3480,3-fluorobenzoic acid,ksc235q1b,paragos 390192
9968
CHEBI:20021
3-fluorobenzoic acid
C1=CC(=CC(=C1)F)C(=O)O
Quantity 5g
Infrared Spectrum Authentic
Melting Point 121°C to 125°C
Color White to Yellow
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula FC6H4CO2H
Beilstein 09, 333
Solubility Information Solubility in water: .. Other solubilities: soluble in pyridine
Formula Weight 140.11
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 3-Fluorobenzoic acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-248-0

TSCA : TSCA