Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluorobenzonitrile, 98%

Click to view available options
Quantity:
5g
25g
100g
403-54-3
C7H4FN
121.114
MFCD00001797
JZTPKAROPNTQQV-UHFFFAOYSA-N
benzonitrile, 3-fluoro,m-fluorobenzonitrile,m-cyanofluorobenzene,benzonitrile, m-fluoro,3-fluoro-benzonitrile,3-fluorobenzenecarbonitrile,3-cyanofluorobenzene,1-cyano-3-fluorobenzene,ncr cf,m-fluorobenzonitile
67880
3-fluorobenzonitrile
C1=CC(=CC(=C1)F)C#N
403-54-3
C7H4FN
121.114
MFCD00001797
JZTPKAROPNTQQV-UHFFFAOYSA-N
benzonitrile, 3-fluoro,m-fluorobenzonitrile,m-cyanofluorobenzene,benzonitrile, m-fluoro,3-fluoro-benzonitrile,3-fluorobenzenecarbonitrile,3-cyanofluorobenzene,1-cyano-3-fluorobenzene,ncr cf,m-fluorobenzonitile
67880
3-fluorobenzonitrile
C1=CC(=CC(=C1)F)C#N
Quantity 5g
Boiling Point 183°C
Odor Almond-like
Melting Point -16°C
Flash Point 67°C (152°F)
UN Number UN3276
Beilstein 1858941
Formula Weight 121.11
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzonitrile

GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : TSCA