Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Fluorobenzonitrile, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Odor | Almond-like |
Refractive Index | 1.5055 |
Density | 1.14 |
Flash Point | 67°C (152°F) |
Melting Point | -16°C |
Boiling Point | 183°C |
Quantity | 25g |
UN Number | UN3276 |
Beilstein | 1858941 |
Formula Weight | 121.11 |
Show More |
Safety and Handling
GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 206-963-5
TSCA : Yes
Recommended Storage : Ambient temperatures