Alfa Aesar™ 3-Fluorobenzotrichloride, 98%

Manufacturer: Alfa Aesar™ B2183103

Quantity 1g

 View more versions of this product

Catalog No. AAB2183103

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 401-77-4
    Molecular Formula C7H4Cl3F
    Formula Weight 213.47
    MDL Number MFCD00042073
    Chemical Name or Material 3-Fluorobenzotrichloride

    Specifications

    Formula Weight 213.47
    MDL Number MFCD00042073
    Health Hazard 1 Danger
    Boiling Point 119° to 120°C (45mmHg)
    Molecular Formula C7H4Cl3F
    CAS 401-77-4
    Health Hazard 2 GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Chemical Name or Material 3-Fluorobenzotrichloride
    Assay 98%

    GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit