Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
456-47-3
C7H7FO
126.13
MFCD00004631
QDHRSLFSDGCJFX-UHFFFAOYSA-N
3-fluorobenzyl alcohol,3-fluorophenyl methanol,m-fluorobenzyl alcohol,benzenemethanol, 3-fluoro,3-fluorobenzylalcohol,3-fluorobenzylic alcohol,benzyl alcohol, m-fluoro,3-fluorophenyl methan-1-ol,3-florobenzyl alcohol,pubchem3486
68008
(3-fluorophenyl)methanol
C1=CC(=CC(=C1)F)CO
456-47-3
C7H7FO
126.13
MFCD00004631
QDHRSLFSDGCJFX-UHFFFAOYSA-N
3-fluorobenzyl alcohol,3-fluorophenyl methanol,m-fluorobenzyl alcohol,benzenemethanol, 3-fluoro,3-fluorobenzylalcohol,3-fluorobenzylic alcohol,benzyl alcohol, m-fluoro,3-fluorophenyl methan-1-ol,3-florobenzyl alcohol,pubchem3486
68008
(3-fluorophenyl)methanol
C1=CC(=CC(=C1)F)CO
Flash Point 90°C
Specific Gravity 1.16
Boiling Point 104°C to 105°C (22.0mmHg)
Packaging Glass bottle
Refractive Index 1.5080 to 1.511
Quantity 25g
Infrared Spectrum Authentic
Density 1.1600g/mL
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H4CH2OH
Formula Weight 126.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzyl alcohol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-265-3

TSCA : TSCA