Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzyl Bromide 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
456-41-7
C7H6BrF
189.027
MFCD00000340
SCBZBMXPJYMXRC-UHFFFAOYSA-N
3-fluorobenzyl bromide,1-bromomethyl-3-fluorobenzene,3-fluorobenzylbromide,m-fluorobenzyl bromide,benzene, 1-bromomethyl-3-fluoro,m-fluorobenzylbromide,alpha-bromo-m-fluorotoluene,alpha-bromo-3-fluorotoluene,toluene, .alpha.-bromo-m-fluoro,1-bromobenzyl-3-fluorobenzene
68007
1-(bromomethyl)-3-fluorobenzene
C1=CC(=CC(=C1)F)CBr
456-41-7
C7H6BrF
189.027
MFCD00000340
SCBZBMXPJYMXRC-UHFFFAOYSA-N
3-fluorobenzyl bromide,1-bromomethyl-3-fluorobenzene,3-fluorobenzylbromide,m-fluorobenzyl bromide,benzene, 1-bromomethyl-3-fluoro,m-fluorobenzylbromide,alpha-bromo-m-fluorotoluene,alpha-bromo-3-fluorotoluene,toluene, .alpha.-bromo-m-fluoro,1-bromobenzyl-3-fluorobenzene
68007
1-(bromomethyl)-3-fluorobenzene
C1=CC(=CC(=C1)F)CBr
Density 1.541g/mL
Color Colorless to Yellow
Packaging Glass bottle
Refractive Index 1.5450 to 1.5480 (20°C, 589nm)
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula FC6H4CH2Br
Beilstein 05, II, 238
Formula Weight 189.03
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 3-Fluorobenzyl bromide, 95%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-263-2

TSCA : TSCA