Alfa Aesar™ 3-Fluorocatechol, 99%

Manufacturer: Alfa Aesar™ B2214503

Quantity 1g

 View more versions of this product

Catalog No. AAB2214503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 363-52-0
    Molecular Formula C6H5FO2
    Formula Weight 128.1
    MDL Number MFCD00042582
    UN Number UN1759
    Beilstein 2436074
    Synonym 1,2-Dihydroxy-3-fluorobenzene
    Chemical Name or Material 3-Fluorocatechol

    Specifications

    Formula Weight 128.1
    Molecular Formula C6H5FO2
    UN Number UN1759
    Beilstein 2436074
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Melting Point 71° to 73°C
    Quantity 1g
    MDL Number MFCD00042582
    CAS 363-52-0
    Synonym 1,2-Dihydroxy-3-fluorobenzene
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Chemical Name or Material 3-Fluorocatechol
    Assay 99%

    GHS H Statement
    H314-H318-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit