Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenethylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
404-70-6
C8H10FN
139.17
MFCD00075376
AUCVZEYHEFAWHO-UHFFFAOYSA-N
3-fluorophenethylamine,2-3-fluorophenyl ethylamine,2-3-fluoro-phenyl-ethylamine,2-3-fluorophenyl ethanamine,2-3-fluorophenyl ethan-1-amine,benzeneethanamine, 3-fluoro,m-fluorophenethylamine,3-flourophenethylamine,m-fluorophenylethylamine,3-fluoro phenethylamine
533928
2-(3-fluorophenyl)ethanamine
C1=CC(=CC(=C1)F)CCN
404-70-6
C8H10FN
139.17
MFCD00075376
AUCVZEYHEFAWHO-UHFFFAOYSA-N
3-fluorophenethylamine,2-3-fluorophenyl ethylamine,2-3-fluoro-phenyl-ethylamine,2-3-fluorophenyl ethanamine,2-3-fluorophenyl ethan-1-amine,benzeneethanamine, 3-fluoro,m-fluorophenethylamine,3-flourophenethylamine,m-fluorophenylethylamine,3-fluoro phenethylamine
533928
2-(3-fluorophenyl)ethanamine
C1=CC(=CC(=C1)F)CCN
Boiling Point 87°C (15.0mmHg)
Flash Point 82°C
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5080 to 1.51
Specific Gravity 1.066
Color Colorless to Yellow
Density 1.0660g/mL
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula FC6H4CH2CH2NH2
Formula Weight 139.17
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3-Fluorophenethylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger