Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
200mL
Packaging:
Glass bottle
372-20-3
C6H5FO
112.1
MFCD00002254
SJTBRFHBXDZMPS-UHFFFAOYSA-N
m-fluorophenol,phenol, 3-fluoro,3-fluoro-phenol,phenol, m-fluoro,meta-fluorophenol,unii-t7oma38487,m-fluoro-phenol,3-fluorphenol,3-fluorophenol,3-fluoro phenol
9743
3-fluorophenol
C1=CC(=CC(=C1)F)O
372-20-3
C6H5FO
112.1
MFCD00002254
SJTBRFHBXDZMPS-UHFFFAOYSA-N
m-fluorophenol,phenol, 3-fluoro,3-fluoro-phenol,phenol, m-fluoro,meta-fluorophenol,unii-t7oma38487,m-fluoro-phenol,3-fluorphenol,3-fluorophenol,3-fluoro phenol
9743
3-fluorophenol
C1=CC(=CC(=C1)F)O
Boiling Point 178.0°C
Specific Gravity 1.24
Infrared Spectrum Authentic
Quantity 200mL
Packaging Glass bottle
Refractive Index 1.5125 to 1.5155
Density 1.2400g/mL
Melting Point 10.0°C
Color Brown-Yellow to Colorless
Flash Point 71°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4OH
Beilstein 06, I, 97
Solubility Information Solubility in water: insoluble
Formula Weight 112.1
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Fluorophenol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : this product darkens during storage