Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. F064325G
Click to view available options
Quantity:
25 g

Specifications

CAS 404-72-8
Color Yellow
Boiling Point 227°C
Molecular Formula C7H4FNS
Quantity 25 g
MDL Number MFCD00004804
UN Number 2922
Synonym 3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
InChI Key HDBACITVPQEAGG-UHFFFAOYSA-N
SMILES FC1=CC=CC(=C1)N=C=S
IUPAC Name 1-fluoro-3-isothiocyanatobenzene
Molecular Weight (g/mol) 153.17
PubChem CID 9834
Formula Weight 153.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorophenyl Isothiocyanate
Show More Show Less
RTECSNumber NX8750000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.