Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluorophenylacetone, 97%

Click to view available options
Quantity:
1g
5g
1737-19-5
C9H9FO
152.168
MFCD00061144
UWCPYXSRCQVABG-UHFFFAOYSA-N
3-fluorophenylacetone,1-3-fluorophenyl propan-2-one,1-3-fluorophenyl acetone,acmc-1bqff,m-fluorophenyl-2-propanone,1-3-fluorophenyl acetone #,1-3-fluorophenyl-2-propanone
137185
1-(3-fluorophenyl)propan-2-one
CC(=O)CC1=CC(=CC=C1)F
1737-19-5
C9H9FO
152.168
MFCD00061144
UWCPYXSRCQVABG-UHFFFAOYSA-N
3-fluorophenylacetone,1-3-fluorophenyl propan-2-one,1-3-fluorophenyl acetone,acmc-1bqff,m-fluorophenyl-2-propanone,1-3-fluorophenyl acetone #,1-3-fluorophenyl-2-propanone
137185
1-(3-fluorophenyl)propan-2-one
CC(=O)CC1=CC(=CC=C1)F
Applications
3-Fluorophenylacetone is used in the preparation of of optically active cyanohydrins. 3-Fluorophenylaceton is reactant in chiral amine synthesis.

Solubility
Soluble in chloroform, ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Quantity 5g
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 3-Fluorophenylacetone