Alfa Aesar™ 3-Fluorophenylmagnesium bromide, 1M in MeTHF

Manufacturer: Alfa Aesar™ H51168AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH51168AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17318-03-5
    Molecular Formula C6H4BrFMg
    Formula Weight 199.31
    MDL Number MFCD00672007
    UN Number UN2924
    Chemical Name or Material 3-Fluorophenylmagnesium bromide

    Specifications

    Formula Weight 199.31
    Molecular Formula C6H4BrFMg
    MDL Number MFCD00672007
    CAS 17318-03-5
    UN Number UN2924
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H302
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Concentration 1M in MeTHF
    Flash Point −11°C (12°F)
    Quantity 500mL
    Chemical Name or Material 3-Fluorophenylmagnesium bromide

    Description

    Applications
    3-Fluorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

    Solubility
    It reacts violently with water.

    Notes
    Air & moisture sensitive. Incompatible with oxidizing agents, acids, bases and alcohols.

    GHS H Statement
    H225-H314-H302
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit