Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoropyridine-2-carboxaldehyde, 98%

Click to view available options
Quantity:
250mg
1g
5g
31224-43-8
C6H4FNO
125.102
MFCD07781234
OZIMPUNGBUYCSP-UHFFFAOYSA-N
3-fluoro-2-formylpyridine,3-fluoropicolinaldehyde,3-fluoro-2-pyridinecarboxaldehyde,3-fluoropyridine-2-carboxaldehyde,3-fluoro-pyridine-2-carbaldehyde,2-pyridinecarboxaldehyde, 3-fluoro,2-formyl-3-fluoropyridine,pubchem5142,acmc-209hkw,3-fluoro-2-formyl-pyridine
11344017
3-fluoropyridine-2-carbaldehyde
C1=CC(=C(N=C1)C=O)F
31224-43-8
C6H4FNO
125.102
MFCD07781234
OZIMPUNGBUYCSP-UHFFFAOYSA-N
3-fluoro-2-formylpyridine,3-fluoropicolinaldehyde,3-fluoro-2-pyridinecarboxaldehyde,3-fluoropyridine-2-carboxaldehyde,3-fluoro-pyridine-2-carbaldehyde,2-pyridinecarboxaldehyde, 3-fluoro,2-formyl-3-fluoropyridine,pubchem5142,acmc-209hkw,3-fluoro-2-formyl-pyridine
11344017
3-fluoropyridine-2-carbaldehyde
C1=CC(=C(N=C1)C=O)F
Applications
3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Quantity 250mg
Solubility Information Slightly soluble in water.
Formula Weight 125.1
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine-2-carboxaldehyde

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures