Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoropyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
372-47-4
C5H4FN
97.09
MFCD00006374
CELKOWQJPVJKIL-UHFFFAOYSA-N
pyridine, 3-fluoro,3-fluorpyridin,ccris 1717,3-fluropyridine,3-fluoro-pyridine,3-fluoro-pyridinium,pubchem1095,acmc-1coty,3-fluoropyridine,ksc173o1l
67794
3-fluoropyridine
C1=CC(=CN=C1)F
372-47-4
C5H4FN
97.09
MFCD00006374
CELKOWQJPVJKIL-UHFFFAOYSA-N
pyridine, 3-fluoro,3-fluorpyridin,ccris 1717,3-fluropyridine,3-fluoro-pyridine,3-fluoro-pyridinium,pubchem1095,acmc-1coty,3-fluoropyridine,ksc173o1l
67794
3-fluoropyridine
C1=CC(=CN=C1)F
Refractive Index 1.4720 to 1.4740
Packaging Glass bottle
Specific Gravity 1.13
Flash Point 13°C
Density 1.1300g/mL
Quantity 1g
Infrared Spectrum Authentic
Boiling Point 107.0°C to 108.0°C
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 20, IV, 2492
Formula Weight 97.09
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Fluoropyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or

GHS Signal Word: Danger

EINECSNumber : 206-755-4