Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoropyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
372-47-4
C5H4FN
97.092
MFCD00006374
CELKOWQJPVJKIL-UHFFFAOYSA-N
pyridine, 3-fluoro,3-fluorpyridin,ccris 1717,3-fluropyridine,3-fluoro-pyridine,3-fluoro-pyridinium,pubchem1095,acmc-1coty,3-fluoropyridine,ksc173o1l
67794
3-fluoropyridine
C1=CC(=CN=C1)F
372-47-4
C5H4FN
97.092
MFCD00006374
CELKOWQJPVJKIL-UHFFFAOYSA-N
pyridine, 3-fluoro,3-fluorpyridin,ccris 1717,3-fluropyridine,3-fluoro-pyridine,3-fluoro-pyridinium,pubchem1095,acmc-1coty,3-fluoropyridine,ksc173o1l
67794
3-fluoropyridine
C1=CC(=CN=C1)F
Flash Point 13°C
Refractive Index 1.4720 to 1.4740
Packaging Glass bottle
Boiling Point 107.0°C to 108.0°C
Infrared Spectrum Authentic
Density 1.1300g/mL
Quantity 5g
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, IV, 2492
Formula Weight 97.09
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Fluoropyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger