Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluorothiophenol, 98%

Click to view available options
Quantity:
1g
5g
25g
2557-77-9
C6H5FS
128.164
MFCD00040227
ZDEUGINAVLMAET-UHFFFAOYSA-N
3-fluorothiophenol,m-fluorothiophenol,m-fluorobenzenethiol,3-fluoro thiophenol,benzenethiol, 3-fluoro,3-fluorobenzene-1-thiol,1-fluoro-3-mercaptobenzene,3-fluoro-thiophenol,3-fluoro benzenethiol,3-fluoro-benzenethiol
75710
3-fluorobenzenethiol
C1=CC(=CC(=C1)S)F
2557-77-9
C6H5FS
128.164
MFCD00040227
ZDEUGINAVLMAET-UHFFFAOYSA-N
3-fluorothiophenol,m-fluorothiophenol,m-fluorobenzenethiol,3-fluoro thiophenol,benzenethiol, 3-fluoro,3-fluorobenzene-1-thiol,1-fluoro-3-mercaptobenzene,3-fluoro-thiophenol,3-fluoro benzenethiol,3-fluoro-benzenethiol
75710
3-fluorobenzenethiol
C1=CC(=CC(=C1)S)F
Odor Stench
Boiling Point 156°C
Flash Point 58°C (136°F)
Quantity 25g
UN Number UN3071
Beilstein 2039302
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 3-Fluorothiophenol

GHS H Statement
H300-H310-H331-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, TOXIC, FLAMMABLE, N.O.S.