Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Formyl-6-methylchromone, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
42059-81-4
C11H8O3
188.182
MFCD00138943
GBWMIOYSMWCYIZ-UHFFFAOYSA-N
3-formyl-6-methylchromone,6-methyl-4-oxo-4h-chromene-3-carbaldehyde,6-methylchromone-3-carboxaldehyde,6-methyl-4-oxo-4h-1-benzopyran-3-carboxaldehyde,4h-1-benzopyran-3-carboxaldehyde, 6-methyl-4-oxo,acmc-1ao5l,methyloxochromenecarbaldehyde,6-methyl-4-oxo-chromene-3-carbaldehyde,4h-1-benzopyran-3-carboxaldehyde,6-methyl-4-oxo
688709
6-methyl-4-oxochromene-3-carbaldehyde
CC1=CC2=C(C=C1)OC=C(C2=O)C=O
42059-81-4
C11H8O3
188.182
MFCD00138943
GBWMIOYSMWCYIZ-UHFFFAOYSA-N
3-formyl-6-methylchromone,6-methyl-4-oxo-4h-chromene-3-carbaldehyde,6-methylchromone-3-carboxaldehyde,6-methyl-4-oxo-4h-1-benzopyran-3-carboxaldehyde,4h-1-benzopyran-3-carboxaldehyde, 6-methyl-4-oxo,acmc-1ao5l,methyloxochromenecarbaldehyde,6-methyl-4-oxo-chromene-3-carbaldehyde,4h-1-benzopyran-3-carboxaldehyde,6-methyl-4-oxo
688709
6-methyl-4-oxochromene-3-carbaldehyde
CC1=CC2=C(C=C1)OC=C(C2=O)C=O
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 169°C to 174°C
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 188.18
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Formyl-6-methylchromone
Show More Show Less