Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Formylsalicylic acid hydrate, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Melting Point | 249°C to 252°C |
Quantity | 5g |
Packaging | Glass bottle |
Assay Percent Range | 96% min. (HPLC) |
Formula Weight | 166.13 |
Percent Purity | 97% |
Chemical Name or Material | 3-Formylsalicylic acid hydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning