Alfa Aesar™ 3-Furaldehyde, 97%, stab. with »0.05% BHT

Manufacturer: Alfa Aesar™ A1133414

Quantity 25g

 View more versions of this product

Catalog No. AAA1133414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 498-60-2
    Molecular Formula C5H4O2
    Formula Weight 96.09
    MDL Number MFCD00010424
    UN Number UN1199
    Beilstein 105852
    Synonym Furan-3-carboxaldehyde
    Chemical Name or Material 3-Furaldehyde, Stabilized with ≈0.05% BHT

    Specifications

    Formula Weight 96.09
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C5H4O2
    Synonym Furan-3-carboxaldehyde
    MDL Number MFCD00010424
    CAS 498-60-2
    UN Number UN1199
    Beilstein 105852
    Quantity 25g
    Flash Point 48°C (118°F)
    Recommended Storage 2°C to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: FURALDEHYDES
    Health Hazard 1 Danger
    Boiling Point 145°C
    Health Hazard 2 GHS H Statement
    H300-H330-H226-H315-H319-H335
    Fatal if swallowed.
    Fatal if inhaled.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Almond-like
    Chemical Name or Material 3-Furaldehyde, Stabilized with ≈0.05% BHT
    Assay 97%

    Description

    Applications
    3-Furaldehyde is compound of honey of various floral origin. It is used to produce N-furan-3-ylmethylene-N ft.-pyridin-2-yl-hydrazine by reaction with 2-hydrazino-pyridine. It is also used as a solvent.

    Solubility
    Soluble in water, benzene, chloroform, alcohol and ether.

    Notes
    Air & Light Sensitive. Store at 4°C. Store in the dark. Incompatible with bases, light and air.

    GHS H Statement
    H300-H330-H226-H315-H319-H335
    Fatal if swallowed.
    Fatal if inhaled.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit