Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Glycidoxypropyltrimethoxysilane, 97%, Thermo Scientific™
Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Density | 1.0700g/mL |
Color | Undesignated |
Boiling Point | 120.0°C (2.0 mmHg) |
Flash Point | 122°C |
Assay Percent Range | 96% min. (GC) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Beilstein | 17, V,3, 45 |
Refractive Index | 1.4280 to 1.4300 |
Quantity | 5g |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 219-784-2
RUO – Research Use Only