Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Heptanol, 98%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
589-82-2
C7H16O
116.204
RZKSECIXORKHQS-UHFFFAOYSA-N
3-heptanol,3-hydroxyheptane,ethyl butyl carbinol,butyl ethyl carbinol,butylethylcarbinol,xi-3-heptanol,fema no. 3547,ethylbutylcarbinol,heptanol-3,3-heptyl alcohol
11520
heptan-3-ol
CCCCC(CC)O
589-82-2
C7H16O
116.204
RZKSECIXORKHQS-UHFFFAOYSA-N
3-heptanol,3-hydroxyheptane,ethyl butyl carbinol,butyl ethyl carbinol,butylethylcarbinol,xi-3-heptanol,fema no. 3547,ethylbutylcarbinol,heptanol-3,3-heptyl alcohol
11520
heptan-3-ol
CCCCC(CC)O
Quantity 25g
Density 0.8180g/mL
Specific Gravity 0.818
Boiling Point 156.0°C
Refractive Index 1.4214
Infrared Spectrum Authentic
Flash Point 55°C
Linear Formula CH3(CH2)3CH(OH)C2H5
Solubility Information Solubility in water: insoluble.
Formula Weight 116.2
Percent Purity 98%
Chemical Name or Material 3-Heptanol
Show More Show Less

GHS H Statement Flammable liquid and vapour. Harmful if swallowed.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Store in a well-ventilated place. Keep container tightly closed.

Warning