Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Heptene, cis + trans, 94%

Click to view available options
Quantity:
5g
25g
100g
592-78-9
C7H14
98.189
MFCD00009475
WZHKDGJSXCTSCK-FNORWQNLSA-N
trans-3-heptene,e-hept-3-ene,e-3-heptene,3-heptene, e,3-heptene, 3e,unii-o4br0a824i,hept-3-ene,cis-3-heptene,3-heptene c,t,z-hept-3-ene
5357259
(E)-hept-3-ene
CCCC=CCC
592-78-9
C7H14
98.189
MFCD00009475
WZHKDGJSXCTSCK-FNORWQNLSA-N
trans-3-heptene,e-hept-3-ene,e-3-heptene,3-heptene, e,3-heptene, 3e,unii-o4br0a824i,hept-3-ene,cis-3-heptene,3-heptene c,t,z-hept-3-ene
5357259
(E)-hept-3-ene
CCCC=CCC
Quantity 100g
Boiling Point 95°C to 96°C
UN Number UN3295
Formula Weight 98.19
Percent Purity 94%
Chemical Name or Material 3-Heptene, cis + trans

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.