Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexanone, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
589-38-8
C6H12O
100.16
MFCD00009402
PFCHFHIRKBAQGU-UHFFFAOYSA-N
3-hexanone,ethyl propyl ketone,3-oxohexane,aethylpropylketon,ethyl n-propyl ketone,3-hexanone natural,aethylpropylketon german,fema no. 3290,3-hexanone, analytical standard,pentanone, methyl
11509
hexan-3-one
CCCC(=O)CC
589-38-8
C6H12O
100.16
MFCD00009402
PFCHFHIRKBAQGU-UHFFFAOYSA-N
3-hexanone,ethyl propyl ketone,3-oxohexane,aethylpropylketon,ethyl n-propyl ketone,3-hexanone natural,aethylpropylketon german,fema no. 3290,3-hexanone, analytical standard,pentanone, methyl
11509
hexan-3-one
CCCC(=O)CC
Boiling Point 123°C
Flash Point 14°C
Specific Gravity 0.81
Refractive Index 1.3990 to 1.401
Density 0.8100g/mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 10mL
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2CH2COC2H5
Beilstein 01, 690
Formula Weight 100.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3-Hexanone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger