Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexyn-2-one, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1679-36-3
C6H8O
96.129
MFCD00041627
LTAPKZGQTMVYMX-UHFFFAOYSA-N
3-hexyn-2-one,2-oxo-3-hexyne,acmc-209dxd,1-butynyl methyl ketone
137151
hex-3-yn-2-one
CCC#CC(=O)C
1679-36-3
C6H8O
96.129
MFCD00041627
LTAPKZGQTMVYMX-UHFFFAOYSA-N
3-hexyn-2-one,2-oxo-3-hexyne,acmc-209dxd,1-butynyl methyl ketone
137151
hex-3-yn-2-one
CCC#CC(=O)C
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.878
Density 0.8780g/mL
Quantity 5g
Color Yellow
Flash Point 39°C
Boiling Point 68.0°C (50.0 mmHg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CH2C≡CCOCH3
Formula Weight 96.13
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Hexyn-2-one
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Fatal if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger