Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-1-methyl-5,6-indolinedione, 98%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
54-06-8
C9H9NO3
179.17
MFCD00069732
RPHLQSHHTJORHI-UHFFFAOYSA-N
adrenochrome, adraxone, usaf uctl-7, 3-hydroxy-1-methyl-5,6-indolinedione, 1-adrenochrome, 3-hydroxy-1-methylindoline-5,6-dione, 2,3-dihydro-3-hydroxy-1-methyl-1h-indole-5,6-dione, 5,6-indolinedione, 3-hydroxy-1-methyl, adrenochrome, l, adrenochrom
5898
3-hydroxy-1-methyl-2,3-dihydroindole-5,6-dione
CN1CC(C2=CC(=O)C(=O)C=C21)O
54-06-8
C9H9NO3
179.17
MFCD00069732
RPHLQSHHTJORHI-UHFFFAOYSA-N
adrenochrome, adraxone, usaf uctl-7, 3-hydroxy-1-methyl-5,6-indolinedione, 1-adrenochrome, 3-hydroxy-1-methylindoline-5,6-dione, 2,3-dihydro-3-hydroxy-1-methyl-1h-indole-5,6-dione, 5,6-indolinedione, 3-hydroxy-1-methyl, adrenochrome, l, adrenochrom
5898
3-hydroxy-1-methyl-2,3-dihydroindole-5,6-dione
CN1CC(C2=CC(=O)C(=O)C=C21)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Merck Index 15, 161
Quantity 250mg
Formula Weight 179.17
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-1-methyl-5, 6-indolinedione

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-192-8

RUO – Research Use Only