Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-1-methylpiperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
3554-74-3
C6H13NO
115.17
UKANCZCEGQDKGF-UHFFFAOYSA-N
3-hydroxy-1-methylpiperidine,1-methyl-3-piperidinol,n-methyl-3-piperidinol,3-piperidinol, 1-methyl,3-hydroxy-n-methylpiperidine,1-methyl-3-hydroxypiperidine,n-methyl-3-hydroxypiperidine,n-methyl-3-hydroxy piperidine,3r-1-methyl-3-piperidinol,pubchem6759
98016
1-methylpiperidin-3-ol
CN1CCCC(C1)O
3554-74-3
C6H13NO
115.17
UKANCZCEGQDKGF-UHFFFAOYSA-N
3-hydroxy-1-methylpiperidine,1-methyl-3-piperidinol,n-methyl-3-piperidinol,3-piperidinol, 1-methyl,3-hydroxy-n-methylpiperidine,1-methyl-3-hydroxypiperidine,n-methyl-3-hydroxypiperidine,n-methyl-3-hydroxy piperidine,3r-1-methyl-3-piperidinol,pubchem6759
98016
1-methylpiperidin-3-ol
CN1CCCC(C1)O
Packaging Glass bottle
Density 0.9990g/mL
Quantity 100g
Specific Gravity 0.999
Refractive Index 1.474
Flash Point 70°C
Boiling Point 76.0°C to 78.0°C (11.0mmHg)
Assay Percent Range 97%
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-1-methylpiperidine
Show More Show Less