Alfa Aesar™ 3-Hydroxy-1-methylpyrrolidine, 97%

Manufacturer: Alfa Aesar™ H2812206

Quantity 5g

 View more versions of this product

Catalog No. AAH2812206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13220-33-2
    Molecular Formula C5H11NO
    Formula Weight 101.15
    MDL Number MFCD00003176
    Synonym 1-Methyl-3-pyrrolidinol
    Chemical Name or Material 3-Hydroxy-1-methylpyrrolidine

    Specifications

    Formula Weight 101.15
    Synonym 1-Methyl-3-pyrrolidinol
    Quantity 5g
    Molecular Formula C5H11NO
    MDL Number MFCD00003176
    CAS 13220-33-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 71°C (160°F)
    Boiling Point 50° to 52°C (1mmHg)
    Chemical Name or Material 3-Hydroxy-1-methylpyrrolidine
    Assay 97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit