Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2,4,6-tribromobenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
14348-40-4
C7H3Br3O3
374.84
MFCD00055557
YDBHVMTTYXWHLI-UHFFFAOYSA-N
3-hydroxy-2,4,6-tribromobenzoic acid,tbhba,benzoic acid, 2,4,6-tribromo-3-hydroxy,2,4,6-tribromo-3-hydroxybenzoicacid,2,4,6-thba,pubchem12541,acmc-209uep,ksc174o3h,2,4,6-tribromo-3-hydroxy-benzoic acid,2,4,6-tribromo-3-hydroxybenzoic acid
151915
2,4,6-tribromo-3-hydroxybenzoic acid
C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br
14348-40-4
C7H3Br3O3
374.84
MFCD00055557
YDBHVMTTYXWHLI-UHFFFAOYSA-N
3-hydroxy-2,4,6-tribromobenzoic acid,tbhba,benzoic acid, 2,4,6-tribromo-3-hydroxy,2,4,6-tribromo-3-hydroxybenzoicacid,2,4,6-thba,pubchem12541,acmc-209uep,ksc174o3h,2,4,6-tribromo-3-hydroxy-benzoic acid,2,4,6-tribromo-3-hydroxybenzoic acid
151915
2,4,6-tribromo-3-hydroxybenzoic acid
C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br
Color White
Melting Point 144°C to 149°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6HBr3(OH)COOH
Solubility Information Solubility in water: insoluble
Formula Weight 374.84
Physical Form Crystals
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2, 4, 6-tribromobenzoic acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA