Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2,4,6-triiodobenzoic acid, 97%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
100g
53279-72-4
C7H3I3O3
515.81
MFCD00020305
GIAVHGFPMPSIFI-UHFFFAOYSA-N
htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
96627
3-hydroxy-2,4,6-triiodobenzoic acid
OC(=O)C1=C(I)C=C(I)C(O)=C1I
53279-72-4
C7H3I3O3
515.81
MFCD00020305
GIAVHGFPMPSIFI-UHFFFAOYSA-N
htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
96627
3-hydroxy-2,4,6-triiodobenzoic acid
OC(=O)C1=C(I)C=C(I)C(O)=C1I
Melting Point 172.5°C to 174.5°C
Assay Percent Range 96% min. (HPLC)
Infrared Spectrum Authentic
Quantity 100g
Formula Weight 515.81
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2,4,6-triiodobenzoic acid

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

EINECSNumber : 258-457-9

RUO – Research Use Only