Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Hydroxy-2,4,6-triiodobenzoic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
53279-72-4
C7H3I3O3
515.811
MFCD00020305
GIAVHGFPMPSIFI-UHFFFAOYSA-N
htba,2,4,6-triiodo-3-hydroxybenzoic acid,benzoic acid, 3-hydroxy-2,4,6-triiodo,3-hydroxy-2,4,6-triiodobenzoicacid,pubchem17410,acmc-1aozs,ksc496i6r,3 hydroxy-2,4,6-triiodobenzoic acid,3-hydroxy-2,4,6 triiodobenzoic acid
96627
3-hydroxy-2,4,6-triiodobenzoic acid
C1=C(C(=C(C(=C1I)O)I)C(=O)O)I
53279-72-4
C7H3I3O3
515.811
MFCD00020305
GIAVHGFPMPSIFI-UHFFFAOYSA-N
htba,2,4,6-triiodo-3-hydroxybenzoic acid,benzoic acid, 3-hydroxy-2,4,6-triiodo,3-hydroxy-2,4,6-triiodobenzoicacid,pubchem17410,acmc-1aozs,ksc496i6r,3 hydroxy-2,4,6-triiodobenzoic acid,3-hydroxy-2,4,6 triiodobenzoic acid
96627
3-hydroxy-2,4,6-triiodobenzoic acid
C1=C(C(=C(C(=C1I)O)I)C(=O)O)I
Melting Point 211°C to 215°C
Quantity 5g
Beilstein 2113494
Formula Weight 515.81
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2,4,6-triiodobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning