Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride, 85%, Acros Organics™

Click to view available options
Quantity:
5g
25g
14173-30-9
C6H8ClNO2
161.585
SKVRYQUMKJQZFN-UHFFFAOYSA-N
3-hydroxy-2-pyridinemethanol hydrochloride,2-hydroxymethyl pyridin-3-ol hydrochloride,3-hydroxy-2-hydroxymethyl pyridine hydrochloride,2-pyridinemethanol, 3-hydroxy-, hydrochloride,2-hydroxymethyl pyridin-3-ol hcl,3-hydroxy-2-hydroxymethylpyridine hydrochloride,pubchem4075,3-hydroxy-2-hydroxymethyl pyridine hcl,acmc-209xde,3-hydroxy-2-pyridinemethanol hcl
3084185
2-(hydroxymethyl)pyridin-3-ol;hydrochloride
C1=CC(=C(N=C1)CO)O.Cl
14173-30-9
C6H8ClNO2
161.585
SKVRYQUMKJQZFN-UHFFFAOYSA-N
3-hydroxy-2-pyridinemethanol hydrochloride,2-hydroxymethyl pyridin-3-ol hydrochloride,3-hydroxy-2-hydroxymethyl pyridine hydrochloride,2-pyridinemethanol, 3-hydroxy-, hydrochloride,2-hydroxymethyl pyridin-3-ol hcl,3-hydroxy-2-hydroxymethylpyridine hydrochloride,pubchem4075,3-hydroxy-2-hydroxymethyl pyridine hcl,acmc-209xde,3-hydroxy-2-pyridinemethanol hcl
3084185
2-(hydroxymethyl)pyridin-3-ol;hydrochloride
C1=CC(=C(N=C1)CO)O.Cl
Quantity 25g
Infrared Spectrum Authentic
Melting Point 217.0°C
Linear Formula HO(C5H3N)CH2OH
Formula Weight 161.59
Percent Purity ≥84.0% (HPLC)
Grade Technical
Chemical Name or Material 3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride

GHS H Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

Warning