Alfa Aesar™ 3-Hydroxy-2-methyl-4-pyrone, 99%

Manufacturer: Alfa Aesar™ A1073514

Quantity 25g

 View more versions of this product

Catalog No. AAA1073514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 118-71-8
    Molecular Formula C6H6O3
    Formula Weight 126.11
    MDL Number MFCD00006578
    Beilstein 112169
    Merck Index 14,5713
    Synonym Maltol
    Chemical Name or Material 3-Hydroxy-2-methyl-4-pyrone

    Specifications

    Formula Weight 126.11
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H6O3
    CAS 118-71-8
    Merck Index 14,5713
    MDL Number MFCD00006578
    Beilstein 112169
    Synonym Maltol
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 95°C (203°F)
    Melting Point 161° to 165°C
    Quantity 25g
    TSCA TSCA
    Health Hazard 1 Danger
    RTECS Number UQ1050000
    Boiling Point 170°C (100mmHg)
    Chemical Name or Material 3-Hydroxy-2-methyl-4-pyrone
    Assay 99%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit