Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2-methyl-4-pyrone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
118-71-8
C6H6O3
126.11
MFCD00006578
XPCTZQVDEJYUGT-UHFFFAOYSA-N
maltol,3-hydroxy-2-methyl-4-pyrone,3-hydroxy-2-methyl-4h-pyran-4-one,larixinic acid,talmon,palatone,vetol,larixic acid,corps praline,4h-pyran-4-one, 3-hydroxy-2-methyl
8369
CHEBI:69438
3-hydroxy-2-methylpyran-4-one
CC1=C(C(=O)C=CO1)O
118-71-8
C6H6O3
126.11
MFCD00006578
XPCTZQVDEJYUGT-UHFFFAOYSA-N
maltol,3-hydroxy-2-methyl-4-pyrone,3-hydroxy-2-methyl-4h-pyran-4-one,larixinic acid,talmon,palatone,vetol,larixic acid,corps praline,4h-pyran-4-one, 3-hydroxy-2-methyl
8369
CHEBI:69438
3-hydroxy-2-methylpyran-4-one
CC1=C(C(=O)C=CO1)O
Boiling Point 170°C (100.0mmHg)
Infrared Spectrum Authentic
Color Colorless
Water 0.5% Max.
Quantity 500g
Packaging Glass bottle
Sulfated Ash 0.2% max.
Melting Point 160°C to 164°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5 % min. (GC)
Merck Index 15, 5777
Solubility Information Solubility in water: 12g/L (25°C). Other solubilities: soluble in alcohol and propylene glycol,sparingly soluble in benzene and ether
Formula Weight 126.11
Physical Form Liquid
Percent Purity ≥98.5% (GC)
Chemical Name or Material 3-Hydroxy-2-methyl-4-pyrone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.