Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2-naphthoic Acid Hydrazide 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5341-58-2
C11H10N2O2
202.213
MFCD00004097
FDNAQCWUERCJBK-UHFFFAOYSA-N
3-hydroxy-2-naphthohydrazide,3-hydroxy-2-naphthoic acid hydrazide,2-hydroxy-3-naphthoic acid hydrazide,2-naphthalenecarboxylic acid, 3-hydroxy-, hydrazide,3-hydroxy-2-naphthoylhydrazine,2-hydroxy-3-naphthoylhydrazine,2-naphthoic acid, 3-hydroxy-, hydrazide,3-hydroxy-2-naphthoic hydrazide,beta-hydroxynaphthoic hydrazide,3-???naphthalen-2-ol
72699
3-hydroxynaphthalene-2-carbohydrazide
C1=CC=C2C=C(C(=CC2=C1)C(=O)NN)O
5341-58-2
C11H10N2O2
202.213
MFCD00004097
FDNAQCWUERCJBK-UHFFFAOYSA-N
3-hydroxy-2-naphthohydrazide,3-hydroxy-2-naphthoic acid hydrazide,2-hydroxy-3-naphthoic acid hydrazide,2-naphthalenecarboxylic acid, 3-hydroxy-, hydrazide,3-hydroxy-2-naphthoylhydrazine,2-hydroxy-3-naphthoylhydrazine,2-naphthoic acid, 3-hydroxy-, hydrazide,3-hydroxy-2-naphthoic hydrazide,beta-hydroxynaphthoic hydrazide,3-???naphthalen-2-ol
72699
3-hydroxynaphthalene-2-carbohydrazide
C1=CC=C2C=C(C(=CC2=C1)C(=O)NN)O
Color Beige
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 206.0°C to 208.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOC10H6CONHNH2
Beilstein 10, 336
Formula Weight 202.21
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide, 97%
Show More Show Less

EINECSNumber : 226-282-7

TSCA : TSCA