Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2-nitropyridine 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
15128-82-2
C5H4N2O3
140.1
MFCD00006258
QBPDSKPWYWIHGA-UHFFFAOYSA-N
3-hydroxy-2-nitropyridine,2-nitropyridin-3-ol,2-nitro-3-hydroxypyridine,2-nitro-3-pyridinol,3-pyridinol, 2-nitro,2-nitro-3-hydroxy pyridine,unii-uu357x85yb,3-hydroxy-2-nitro pyridine,2-nitro-3-pyridol
27057
C1=CC(=C(N=C1)[N+](=O)[O-])O
15128-82-2
C5H4N2O3
140.1
MFCD00006258
QBPDSKPWYWIHGA-UHFFFAOYSA-N
3-hydroxy-2-nitropyridine,2-nitropyridin-3-ol,2-nitro-3-hydroxypyridine,2-nitro-3-pyridinol,3-pyridinol, 2-nitro,2-nitro-3-hydroxy pyridine,unii-uu357x85yb,3-hydroxy-2-nitro pyridine,2-nitro-3-pyridol
27057
C1=CC(=C(N=C1)[N+](=O)[O-])O
Color Yellow
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 67°C to 72°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HO(C5H3N)NO2
Solubility Information Solubility in water: soluble in water
Formula Weight 140.1
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Hydroxy-2-nitropyridine, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 239-191-2

RTECSNumber : UU7715000

TSCA : TSCA