Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methoxybenzoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Plastic bottle
645-08-9
C8H8O4
168.15
MFCD00002507
LBKFGYZQBSGRHY-UHFFFAOYSA-N
isovanillic acid,acide isovanillique,3-hydroxyanisic acid,3-hydroxy-p-anisic acid,benzoic acid, 3-hydroxy-4-methoxy,p-anisic acid, 3-hydroxy,chembl88700,3-hydroxy-4-methoxy-benzoic acid,3-hydroxy-4-methoxybenzoicacid,5-carboxyguaiacol
12575
CHEBI:63798
3-hydroxy-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)O
645-08-9
C8H8O4
168.15
MFCD00002507
LBKFGYZQBSGRHY-UHFFFAOYSA-N
isovanillic acid,acide isovanillique,3-hydroxyanisic acid,3-hydroxy-p-anisic acid,benzoic acid, 3-hydroxy-4-methoxy,p-anisic acid, 3-hydroxy,chembl88700,3-hydroxy-4-methoxy-benzoic acid,3-hydroxy-4-methoxybenzoicacid,5-carboxyguaiacol
12575
CHEBI:63798
3-hydroxy-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)O
Packaging Glass bottle
Color Beige to Brown-Gray
Melting Point 245°C to 250°C
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula HOC6H3(OCH3)CO2H
Beilstein 10, 393
Formula Weight 168.15
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Hydroxy-4-methoxybenzoic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-430-5

RTECSNumber : BZ4850000

TSCA : TSCA