Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methoxycinnamic Acid, Predominantly trans 97%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
537-73-5
C10H10O4
194.186
MFCD00004391
QURCVMIEKCOAJU-HWKANZROSA-N
isoferulic acid,3-hydroxy-4-methoxycinnamic acid,hesperetic acid,3-3-hydroxy-4-methoxyphenyl acrylic acid,2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid,isoferulate,2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl,e-3-3-hydroxy-4-methoxyphenyl acrylic acid,3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid,3-hydroxy-4-methoxy-cinnamic acid
736186
CHEBI:27794
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
COC1=C(C=C(C=C1)C=CC(=O)O)O
537-73-5
C10H10O4
194.186
MFCD00004391
QURCVMIEKCOAJU-HWKANZROSA-N
isoferulic acid,3-hydroxy-4-methoxycinnamic acid,hesperetic acid,3-3-hydroxy-4-methoxyphenyl acrylic acid,2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid,isoferulate,2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl,e-3-3-hydroxy-4-methoxyphenyl acrylic acid,3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid,3-hydroxy-4-methoxy-cinnamic acid
736186
CHEBI:27794
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
COC1=C(C=C(C=C1)C=CC(=O)O)O
Melting Point 228°C to 233°C
Quantity 5g
Packaging Glass bottle
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOC6H3(OCH3)CH=CHCO2H
Beilstein 10, 437
Formula Weight 194.19
Physical Form Powder or Flakes
Percent Purity ≥96.0%
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid, 97%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

EINECSNumber : 208-676-0

RTECSNumber : UD3365400

TSCA : TSCA