Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methoxyphenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
269410-23-3
C13H19BO4
250.1
WINSAMQIAMEZIK-UHFFFAOYSA-N
2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,3-hydroxy-4-methoxyphenylboronic acid, pinacol ester,2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,3-hydroxy-4-methoxyphenylboronic acid pinacol ester,phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,amtb953,2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol
17750251
2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)OC)O
269410-23-3
C13H19BO4
250.1
WINSAMQIAMEZIK-UHFFFAOYSA-N
2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,3-hydroxy-4-methoxyphenylboronic acid, pinacol ester,2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,3-hydroxy-4-methoxyphenylboronic acid pinacol ester,phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,amtb953,2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol
17750251
2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)OC)O
Melting Point 101.0°C to 102.0°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 250.1
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-methoxyphenylboronic acid pinacol ester

GHS Signal Word: Warning