Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-nitrobenzoic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
619-14-7
C7H5NO5
183.12
MFCD00007110
XLDLRRGZWIEEHT-UHFFFAOYSA-N
3-hydroxy-4-nitrobenzoic acid,3-hydroxy-4-nitro-benzoic acid,4-nitro-3-hydroxybenzoic acid,benzoic acid, 3-hydroxy-4-nitro,3-hydroxy-4-nitrobenzoicacid,pubchem17135,acmc-209mxi,3-hydroxy4nitrobenzoic acid,ksc356q8j,3-hydroxy-4-nitro benzoic acid
69265
C1=CC(=C(C=C1C(=O)O)O)[N+](=O)[O-]
619-14-7
C7H5NO5
183.12
MFCD00007110
XLDLRRGZWIEEHT-UHFFFAOYSA-N
3-hydroxy-4-nitrobenzoic acid,3-hydroxy-4-nitro-benzoic acid,4-nitro-3-hydroxybenzoic acid,benzoic acid, 3-hydroxy-4-nitro,3-hydroxy-4-nitrobenzoicacid,pubchem17135,acmc-209mxi,3-hydroxy4nitrobenzoic acid,ksc356q8j,3-hydroxy-4-nitro benzoic acid
69265
C1=CC(=C(C=C1C(=O)O)O)[N+](=O)[O-]
Melting Point 233°C
Quantity 100g
Water 0.5% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Color Brown to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H3(NO2)CO2H
Beilstein 10, 146
Solubility Information Solubility in water: insoluble
Formula Weight 183.12
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Hydroxy-4-nitrobenzoic acid, 98%
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 210-580-9

TSCA : TSCA