Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-quinolinecarboxylic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
118-13-8
C10H7NO3
189.17
MXNVEJDRXSFZQB-UHFFFAOYSA-N
3-hydroxy-4-quinolinecarboxylic acid,3-hydroxycinchoninic acid,cinchoninic acid, 3-hydroxy,3-hydroxy-quinoline-4-carboxylic acid,4-quinolinecarboxylic acid, 3-hydroxy,3-hydroxy-4-quinolinecarboxylicacid,3-hydroxycinchoninicacid,4-22-00-02296 beilstein handbook reference,4-quinolinecarboxylicacid,3-hydroxy,4-quinolinecarboxylicacid, 3-hydroxy
8352
3-hydroxyquinoline-4-carboxylic acid
C1=CC=C2C(=C1)C(=C(C=N2)O)C(=O)O
118-13-8
C10H7NO3
189.17
MXNVEJDRXSFZQB-UHFFFAOYSA-N
3-hydroxy-4-quinolinecarboxylic acid,3-hydroxycinchoninic acid,cinchoninic acid, 3-hydroxy,3-hydroxy-quinoline-4-carboxylic acid,4-quinolinecarboxylic acid, 3-hydroxy,3-hydroxy-4-quinolinecarboxylicacid,3-hydroxycinchoninicacid,4-22-00-02296 beilstein handbook reference,4-quinolinecarboxylicacid,3-hydroxy,4-quinolinecarboxylicacid, 3-hydroxy
8352
3-hydroxyquinoline-4-carboxylic acid
C1=CC=C2C(=C1)C(=C(C=N2)O)C(=O)O
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-quinolinecarboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 204-236-7