Alfa Aesar™ 3-Hydroxyazetidine hydrochloride, 97%

Manufacturer: Alfa Aesar™ H25842MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25842MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18621-18-6
    Molecular Formula C3H7NO·HCl
    Formula Weight 109.56
    MDL Number MFCD02683887
    Synonym 3-Azetidinol hydrochloride
    Chemical Name or Material 3-Hydroxyazetidine hydrochloride

    Specifications

    Formula Weight 109.56
    Health Hazard 1 Warning
    Melting Point 85° to 90°C
    Quantity 250mg
    Molecular Formula C3H7NO·HCl
    MDL Number MFCD02683887
    Synonym 3-Azetidinol hydrochloride
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 18621-18-6
    Chemical Name or Material 3-Hydroxyazetidine hydrochloride
    Assay 97%

    Description

    Applications
    3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.

    Solubility
    Soluble in water, DMSO, methanol.

    Notes
    Hygroscopic. Store under inert gas. Store away from water/ moisture, oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit