Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxybenzoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
99-06-9
C7H6O3
138.122
MFCD00002506
IJFXRHURBJZNAO-UHFFFAOYSA-N
m-hydroxybenzoic acid,m-salicylic acid,3-carboxyphenol,benzoic acid, 3-hydroxy,m-hba,3-hydroxybenzoate,benzoic acid, m-hydroxy,acido m-idrossibenzoico,meta-hydroxybenzoic acid,kyselina 3-hydroxybenzoova
7420
CHEBI:30764
3-hydroxybenzoic acid
C1=CC(=CC(=C1)O)C(=O)O
99-06-9
C7H6O3
138.122
MFCD00002506
IJFXRHURBJZNAO-UHFFFAOYSA-N
m-hydroxybenzoic acid,m-salicylic acid,3-carboxyphenol,benzoic acid, 3-hydroxy,m-hba,3-hydroxybenzoate,benzoic acid, m-hydroxy,acido m-idrossibenzoico,meta-hydroxybenzoic acid,kyselina 3-hydroxybenzoova
7420
CHEBI:30764
3-hydroxybenzoic acid
C1=CC(=CC(=C1)O)C(=O)O
Flash Point 220°C
Quantity 2.5kg
Infrared Spectrum Authentic
Melting Point 201.0°C to 204.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4CO2H
Beilstein 10, 134
Formula Weight 138.12
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Hydroxybenzoic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-726-5

RTECSNumber : DH1924980

TSCA : TSCA