Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Hydroxybenzophenone, 99%

Click to view available options
Quantity:
1g
5g
25g
13020-57-0
C13H10O2
198.221
MFCD00002297
SHULEACXTONYPS-UHFFFAOYSA-N
3-hydroxybenzophenone,3-hydroxyphenyl phenyl methanone,m-hydroxybenzophenone,methanone, 3-hydroxyphenyl phenyl,3-hydroxy-benzophenon,dsstox_cid_27856,dsstox_rid_82609,dsstox_gsid_47880,3-hydroxyphenyl phenyl ketone
83050
(3-hydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O
13020-57-0
C13H10O2
198.221
MFCD00002297
SHULEACXTONYPS-UHFFFAOYSA-N
3-hydroxybenzophenone,3-hydroxyphenyl phenyl methanone,m-hydroxybenzophenone,methanone, 3-hydroxyphenyl phenyl,3-hydroxy-benzophenon,dsstox_cid_27856,dsstox_rid_82609,dsstox_gsid_47880,3-hydroxyphenyl phenyl ketone
83050
(3-hydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O
Applications
3-Hydroxybenzophenone is a benzophenone metabolite with potential estrogenic and anti-androgenic activity. is a more attractive hydrogen bonding co-material for MCPBA, as the 3-hydroxyl group is unable to form an intramolecular hydrogen bond. It is commonly used as a radical and cationic photo initiators.

Solubility
Soluble in Chloroform, DCM. Ethyl Acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 113°C to 115°C
Quantity 25g
Solubility Information Soluble in Chloroform,DCM. Ethyl Acetate.
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material 3-Hydroxybenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-879-1

TSCA : No

Recommended Storage : Ambient temperatures