Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxybenzotrifluoride 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
98-17-9
C7H5F3O
162.111
MFCD00002299
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-trifluoromethyl phenol,3-hydroxybenzotrifluoride,m-trifluoromethylphenol,m-hydroxybenzotrifluoride,phenol, 3-trifluoromethyl,m-trifluoromethyl phenol,alpha,alpha,alpha-trifluoro-m-cresol,a,a,a-trifluoro-m-cresol
7376
3-(trifluoromethyl)phenol
C1=CC(=CC(=C1)O)C(F)(F)F
98-17-9
C7H5F3O
162.111
MFCD00002299
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-trifluoromethyl phenol,3-hydroxybenzotrifluoride,m-trifluoromethylphenol,m-hydroxybenzotrifluoride,phenol, 3-trifluoromethyl,m-trifluoromethyl phenol,alpha,alpha,alpha-trifluoro-m-cresol,a,a,a-trifluoro-m-cresol
7376
3-(trifluoromethyl)phenol
C1=CC(=CC(=C1)O)C(F)(F)F
Boiling Point 178°C
Color Yellow
Quantity 250g
Formula Weight 162.11
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Hydroxybenzotrifluoride

RTECSNumber : GP3510000

TSCA : Yes