Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxydiphenylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
101-18-8
C12H11NO
185.226
NDACNGSDAFKTGE-UHFFFAOYSA-N
3-hydroxydiphenylamine,3-phenylamino phenol,m-anilinophenol,phenol, 3-phenylamino,3-hydroxy-n-phenylaniline,m-hydroxydiphenylamine,3-phenylaminophenol,phenol, m-anilino,unii-e39q946go3,ccris 4658
7546
3-anilinophenol
C1=CC=C(C=C1)NC2=CC(=CC=C2)O
101-18-8
C12H11NO
185.226
NDACNGSDAFKTGE-UHFFFAOYSA-N
3-hydroxydiphenylamine,3-phenylamino phenol,m-anilinophenol,phenol, 3-phenylamino,3-hydroxy-n-phenylaniline,m-hydroxydiphenylamine,3-phenylaminophenol,phenol, m-anilino,unii-e39q946go3,ccris 4658
7546
3-anilinophenol
C1=CC=C(C=C1)NC2=CC(=CC=C2)O
Melting Point 80°C to 84°C
Color Brown
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 185.23
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Hydroxydiphenylamine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning