Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Hydroxymethyl-1-adamantanol, 96%

Click to view available options
Quantity:
250mg
1g
5g
38584-37-1
C11H18O2
182.263
MFCD03105096
FORAJDRXEYKDFJ-UHFFFAOYSA-N
3-hydroxymethyl adamantan-1-ol,3-hydroxymethyl-1-adamantanol,3-hydroxymethyl-1-adamantanol,tricyclo 3.3.1.13,7 decane-1-methanol, 3-hydroxy,3-hydroxymethyl-1-adamantol,3-hydroxy-1-adamantanemethanol,3-hydroxyadamantyl methanol,1-hydroxymethyl-3-adamantanol,3-hydroxy-1-adamantane methanol,1-hydroxy-3-hydroxymethyladamantane
12070475
3-(hydroxymethyl)adamantan-1-ol
C1C2CC3(CC1CC(C2)(C3)O)CO
38584-37-1
C11H18O2
182.263
MFCD03105096
FORAJDRXEYKDFJ-UHFFFAOYSA-N
3-hydroxymethyl adamantan-1-ol,3-hydroxymethyl-1-adamantanol,3-hydroxymethyl-1-adamantanol,tricyclo 3.3.1.13,7 decane-1-methanol, 3-hydroxy,3-hydroxymethyl-1-adamantol,3-hydroxy-1-adamantanemethanol,3-hydroxyadamantyl methanol,1-hydroxymethyl-3-adamantanol,3-hydroxy-1-adamantane methanol,1-hydroxy-3-hydroxymethyladamantane
12070475
3-(hydroxymethyl)adamantan-1-ol
C1C2CC3(CC1CC(C2)(C3)O)CO
Quantity 5g
Color Brown to White
Melting Point 158°C to 162°C
Formula Weight 182.26
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 3-Hydroxymethyl-1-adamantanol